missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Europium Mini Beads Antibody Conjugation Kit

Product Code. 18006204 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
10 Reactions
4 Reactions
Unit Size:
16µg
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18006204 4 Reactions 16µg
18003826 10 Reactions Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18006204 Supplier Novus Biologicals™ Supplier No. 12000040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novus Biologicals™ Europium Mini Antibody Labeling Kit allows antibodies or proteins to quickly and easily conjugated to 200nm Europium chelate microspheres.

Kit Components:
  • 4 or 10 Mini vials of 200nm Europium (dependent on kit)
  • 1 vial of 1 x Reaction Buffer A
  • 1 vial of 1 x Reaction Buffer B
  • 1 vial of 10x Europium Quencher
  • 1 vial of 1 x Resuspension Buffer
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 16 μg
Kit Contents 4 or 10 Mini vials of 200nm Europium (dependent on kit), 1 vial of 1x Reaction Buffer A, 1 vial of 1x Reaction Buffer B, 1 vial of 10x Europium Quencher, 1 vial of 1x Resuspension Buffer
Sufficient For 4 Reactions
For Use With (Application) The optimum amount of antibody (which will influence the number of antibody molecules per particle) depends on the size of the particles (surface area) and on the application. You may need to conjugate different amounts of antibody to optimize your assay. We recommend testing with 0.1 mg/mL in the first instance, although slightly lower or higher concentrations can be explored to optimize performance in your particular application. Before testing antibody concentrations above 0.1 mg/mL we advise you to use our Antibody Concentration & Clean Up Kit for LATEX and EUROPIUM to remove interfering buffer components. This is due to the increase in contamination that will occur when conjugating using a larger volume of the stock antibody. Do not alter the reaction volume of 400 uL as this will reduce conjugation efficiency.
Product Type Europium Mini Antibody Labeling Kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.