missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) thenoyltrifluoroacetonate, trihydrate, 95%, Thermo Scientific Chemicals

Product Code. 10311311
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21392-96-1
Molecular Formula C24H12EuF9O6S3·3H2O
Molecular Weight (g/mol) 869.54
MDL Number MFCD00150912
InChI Key LSMLVRQTTAIDBZ-BSWAEIBTSA-N
Synonym tris 4,4,4-trifluoro-1-2-thienyl-1,3-butanediono europium iii hydrate
PubChem CID 91873327
IUPAC Name erbium;(Z)-1,1,1-trifluoro-4-hydroxy-4-thiophen-2-ylbut-3-en-2-one;hydrate
SMILES C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.O.[Er]

Specifications

Melting Point 142.0°C to 147.0°C
Color Yellow
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 869.54
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Europium(III) thenoyltrifluoroacetonate, trihydrate, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'