missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) sulfate octahydrate, REacton™, 99.99% (REO)

Product Code. 11396658
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396658 25 g 25g
11386658 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396658 Supplier Thermo Scientific Alfa Aesar Supplier No. 013210.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-55-7 | Eu2H16O20S3 | 736.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-55-7
Molecular Formula Eu2H16O20S3
Molecular Weight (g/mol) 736.216
MDL Number MFCD00149704
InChI Key PGDXJFDXKJLVTJ-UHFFFAOYSA-H
Synonym europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8
PubChem CID 25021676
IUPAC Name europium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]

Specifications

Melting Point 375°C (in -8H2O)
Physical Form Crystalline
Quantity 25 g
Merck Index 14,3904
Formula Weight 736.23 (592.10 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) sulfate octahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.