missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) phosphate hydrate, 99.9% (REO)

Product Code. p-7051790
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11364987

Brand: Thermo Scientific Alfa Aesar 041989.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14913-20-3 | EuO4P | 246.93 g/mol

Europium finds application in optical coating and thin film application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-20-3
Molecular Formula EuO4P
Molecular Weight (g/mol) 246.93
MDL Number MFCD05664261
InChI Key KJNZTHUWRRLWOA-UHFFFAOYSA-K
Synonym europium iii phosphate hydrate, europium 3+ hydrate phosphate, europium 3+ phosphate-water 1/1/1, europium 3+ ion hydrate phosphate
PubChem CID 16217374
SMILES [Eu+3].[O-]P([O-])([O-])=O

Specifications

Physical Form Chunks and/or Powder
Quantity 1 g
Linear Formula EuPO4·xH2O
Solubility Information Insoluble in water.
IUPAC Name europium(3+) phosphate
Formula Weight 246.93 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) phosphate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.