missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) oxide, nanopowder, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Europium oxide nanopowder acts as phosphors for lamps. It is used as a fluorescent label in immunoassay for atrazine. It is also used in color TV, X-ray, glass, crystal and other luminescent materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2291°C |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,3904 |
Solubility Information | Insoluble in water. |
Formula Weight | 351.92 |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Assay | (REO) |
Chemical Name or Material | Europium (III) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'