missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11391039
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1308-96-9
Eu2O3
351.93
MFCD00010997
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
159371
europium(3+);oxygen(2-)
[O--].[O--].[O--].[Eu+3].[Eu+3]
This item is not returnable. View return policy
1308-96-9
Eu2O3
351.93
MFCD00010997
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
159371
europium(3+);oxygen(2-)
[O--].[O--].[O--].[Eu+3].[Eu+3]

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2291°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 351.92
Sensitivity Hygroscopic
Appearance White powder or chunks
Density 7.42g/cm3
Chemical Name or Material Europium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'