Learn More
Europium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
For vacuum deposition
£155.00 - £621.00
Chemical Identifiers
CAS | 1308-96-9 |
---|---|
Molecular Formula | Eu2O3 |
Molecular Weight (g/mol) | 351.93 |
MDL Number | MFCD00010997 |
InChI Key | AEBZCFFCDTZXHP-UHFFFAOYSA-N |
Synonym | europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- |
PubChem CID | 159371 |
SMILES | [O--].[O--].[O--].[Eu+3].[Eu+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303448
|
Thermo Scientific Alfa Aesar
036352.04 |
2 g |
£155.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393438
|
Thermo Scientific Alfa Aesar
036352.09 |
10 g | |||||||
Description
Europium(III) oxide is used for vacuum deposition, fluorescent lamps, and as an activator for yttrium-based phosphors, manufacture of fluorescent glass, anti-counterfeiting phosphors in Euro banknotes. And it is widely used as a red or blue phosphor in television sets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-96-9 | |
351.93 | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
159371 |
Eu2O3 | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
[O--].[O--].[O--].[Eu+3].[Eu+3] |
Specifications
2291°C | |
Sintered Lump | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
dieuropium(3+) trioxidandiide | |
159371 | |
99.9% | |
(REO) |
1308-96-9 | |
Eu2O3 | |
14,3904 | |
Soluble in acids. Insoluble in water. | |
[O--].[O--].[O--].[Eu+3].[Eu+3] | |
351.93 | |
351.92 | |
Hygroscopic | |
Europium(III) oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-165-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only