missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxalate hydrate, 99.9% (REO)

Product Code. p-7049586
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11367108

Brand: Thermo Scientific Alfa Aesar 015288.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 152864-32-9 | C6Eu2O12 | 567.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152864-32-9
Molecular Formula C6Eu2O12
Molecular Weight (g/mol) 567.98
MDL Number MFCD00150143
InChI Key PVDYMOCCGHXJAK-UHFFFAOYSA-H
Synonym europium iii oxalate hydrate, c6eu2o12.h2o, ethanedioic acid europium salt hydrate, bis 4,5-dioxo-1,3-dioxa-2-europacyclopentan-2-yl oxalate hydrate
PubChem CID 71310234
IUPAC Name dieuropium(3+) trioxalate
SMILES [Eu+3].[Eu+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Color White
Physical Form Crystalline
Quantity 2 g
Linear Formula Eu2(C2O4)3·xH2O
UN Number UN3288
Formula Weight 567.98 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) oxalate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.