missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) fluoride, anhydrous, 99.5% (REO), Thermo Scientific™

Product Code. 11359209
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359209 1 g 1g
11369209 5 g 5g
11316389 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359209 Supplier Thermo Scientific Alfa Aesar Supplier No. 088181.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-25-8
Molecular Formula EuF3
Molecular Weight (g/mol) 208.96
MDL Number MFCD00010998
InChI Key HPNURIVGONRLQI-UHFFFAOYSA-K
Synonym europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
IUPAC Name europium(3+) trifluoride
SMILES [F-].[F-].[F-].[Eu+3]

Specifications

Boiling Point >2280°C
Melting Point 1390°C
Physical Form Powder
Quantity 1 g
Formula Weight 208.96
Percent Purity 99.5%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium (III) fluoride, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.