missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride hydrate, REacton™, 99.99% (REO)

Product Code. p-4408087
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11311039

Brand: Thermo Scientific Alfa Aesar 011297.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EuCl3·xH2O

Europium(III) chloride hydrate is used as a catalyst in organic synthesis. It is involved in the preparation of amino acid based europium(III) complex in order to prepare photoactive thin films, which finds application in organic light emitting diodes. It is also used as a precursor for the preparation of other europium compounds and its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula EuCl3·xH2O
MDL Number MFCD00010994

Specifications

Color Yellow
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 258.32 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow crystalline
Chemical Name or Material Europium(III) chloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.