missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride hexahydrate, 99.9%, (trace metal basis), -10 mesh

Product Code. 10296490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296490 25 g Glass bottle 25g
10616232 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10296490 Supplier Thermo Scientific Acros Supplier No. 199600250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13759-92-7 | Cl3Eu·6H2O | 366.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13759-92-7
Molecular Formula Cl3Eu·6H2O
Molecular Weight (g/mol) 366.4
MDL Number MFCD00149701

Specifications

Melting Point 850.0°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula EuCl3·6H2O
Merck Index 10, 3854
Solubility Information Solubility in water: soluble
Formula Weight 366.4
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.89
Density 4.8900g/mL
Chemical Name or Material Europium(III) chloride hexahydrate, 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.