missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) bromide hydrate, REacton™, 99.99% (REO)

Product Code. 11346668
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346668 5 g 5g
11336668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346668 Supplier Thermo Scientific Alfa Aesar Supplier No. 013213.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EuBr3·xH2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula EuBr3·xH2O
MDL Number MFCD00049921

Specifications

Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 391.69 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(III) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.