missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) acetate hydrate, REacton™, 99.999% (REO), Thermo Scientific Chemicals

£215.00 - £1085.00

Chemical Identifiers

CAS 62667-64-5
Molecular Formula C6H9EuO6
Molecular Weight (g/mol) 329.10
MDL Number MFCD00150115
InChI Key LNYNHRRKSYMFHF-UHFFFAOYSA-K
Synonym europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis
PubChem CID 16212055
IUPAC Name europium(3+) triacetate
SMILES [Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325128
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044056.09
10 g
£215.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11335128
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044056.18
50 g
£1085.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62667-64-5
329.10
LNYNHRRKSYMFHF-UHFFFAOYSA-K
16212055
[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
C6H9EuO6
MFCD00150115
europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis
europium(3+) triacetate

Specifications

62667-64-5
C6H9EuO6
europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis
[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
329.10
329.10 (Anhydrous)
Reacton™
(REO)
Lump/Powder
MFCD00150115
LNYNHRRKSYMFHF-UHFFFAOYSA-K
europium(3+) triacetate
16212055
99.999%
Hygroscopic
Europium (III) acetate hydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 214-670-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only