missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) acetate hydrate, 99.99%

Product Code. 11489216
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489216 1 g 1g
11499216 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489216 Supplier Thermo Scientific Alfa Aesar Supplier No. A18069.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 62667-64-5 | C6H9EuO6 | 329.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62667-64-5
Molecular Formula C6H9EuO6
Molecular Weight (g/mol) 329.10
MDL Number MFCD00150115
InChI Key LNYNHRRKSYMFHF-UHFFFAOYSA-K
Synonym europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis
PubChem CID 16212055
SMILES [Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Quantity 1 g
Sensitivity Hygroscopic
IUPAC Name acetic acid;europium;hydrate
Formula Weight 329.10 (Anhydrous)
Percent Purity 99.99%
Chemical Name or Material Europium(III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.