missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Product Code. 11376658
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
10g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376658 5 g 5g
11366658 2 g 2g
11356658 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376658 Supplier Thermo Scientific Alfa Aesar Supplier No. 013209.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 181266-82-0 | C15H21EuO6 | 449.29 g/mol

It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181266-82-0
Molecular Formula C15H21EuO6
Molecular Weight (g/mol) 449.29
MDL Number MFCD00192170
InChI Key YMTKZYSHNNXTIJ-UHFFFAOYSA-N
Synonym europium iii 2,4-pentanedionate hydrate reo
PubChem CID 131675702
SMILES [Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 140°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
IUPAC Name europium(3+) tris(2,4-dioxopentan-3-ide)
Formula Weight 449.29 (Anhydrous)
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Europium(III) 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.