missing translation for 'onlineSavingsMsg'
Learn More

Europium(II) bromide, 99.99% (metals basis)

Product Code. 11400730
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11400730

Brand: Thermo Scientific Alfa Aesar 044431.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13780-48-8 | Br2Eu | 311.772 g/mol

Europium(II) bromide is used in water treatment, chemical analysis and in certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13780-48-8
Molecular Formula Br2Eu
Molecular Weight (g/mol) 311.772
MDL Number MFCD01076548
InChI Key PJVPGMOCWCUQHP-UHFFFAOYSA-L
Synonym europium dibromide, europium ii bromide, europium ii ; bromide, acmc-1bp16, europium ii bromide trace rare earth metals basis 1g
PubChem CID 53249294
IUPAC Name europium(2+);dibromide
SMILES [Br-].[Br-].[Eu+2]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 311.77
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(II) bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.