missing translation for 'onlineSavingsMsg'
Learn More

Eupatilin, TRC

Product Code. 30379584
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379584 5 mg
30387367 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379584 Supplier Toronto Research Chemicals Supplier No. TRCE9387005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22368-21-4
Molecular Formula C18 H16 O7
Molecular Weight (g/mol) 344.32
Synonym 4H-1-Benzopyran-4-one, 2-(3,4-dimethoxyphenyl)-5,7-dihydroxy-6-methoxy-, Flavone, 5,7-dihydroxy-3',4',6-trimethoxy- (8CI), 2-(3,4-Dimethoxyphenyl)-5,7-dihydroxy-6-methoxy-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3',4',6-trimethoxyflavone, 5,7-Dihydroxy-6,3',4'-trimethoxyflavone, Eupatilin, NSC 122413, Stillen
IUPAC Name 2-(3,4-dimethoxyphenyl)-5,7-dihydroxy-6-methoxychromen-4-one
SMILES COc1ccc(cc1OC)C2=CC(=O)c3c(O)c(OC)c(O)cc3O2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H16O7/c1-22-12-5-4-9(6-14(12)23-2)13-7-10(19)16-15(25-13)8-11(20)18(24-3)17(16)21/h4-8,20-21H,1-3H3
Formula Weight 344.0896
Percent Purity >95%
Chemical Name or Material Eupatilin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.