missing translation for 'onlineSavingsMsg'
Learn More

Eupatilin, MedChemExpress

Product Code. 16474887
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474887 50 mg 50mg
16459098 10 mg 10mg
16451508 5 mg 5mg
16433906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474887 Supplier Medchem Express Supplier No. HYN0783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eupatilin, a lipophilic flavonoid isolated from Artemisia species, is a PPARα agonist, and possesses anti-apoptotic, anti-oxidative and anti-inflammatory activities.

Eupatilin, a lipophilic flavonoid isolated from Artemisia species, is a PPARα agonist, and possesses anti-apoptotic, anti-oxidative and anti-inflammatory activities.

Specifications

Chemical Name or Material Eupatilin
CAS 22368-21-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H16O7
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (96.80 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=CC(O)=C(OC)C(O)=C13
Molecular Weight (g/mol) 344.32
Formula Weight 344.32
Percent Purity 98.07%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.