missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eukaryotic translation initiation factor 1 Antibody (OTI2B9), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71554MFV500
This item is not returnable.
View return policy
Description
Eukaryotic translation initiation factor 1 Monoclonal antibody specifically detects Eukaryotic translation initiation factor 1 in Human, Rat samples. It is validated for Flow Cytometry
Specifications
| Eukaryotic translation initiation factor 1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Flow Cytometry | |
| OTI2B9 | |
| Flow Cytometry | |
| A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1 | |
| Full length human recombinant protein of human EIF1 (NP_005792) produced in HEK293T cell. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 10209 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction