missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eukaryotic translation initiation factor 1 Antibody (OTI2B9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71554JF646
This item is not returnable.
View return policy
Description
Eukaryotic translation initiation factor 1 Monoclonal specifically detects Eukaryotic translation initiation factor 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Eukaryotic translation initiation factor 1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EIF1 | |
| Full length recombinant protein of human Eukaryotic translation initiation factor 1 (NP_005792) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2B9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 10209.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction