Learn More
Gibertini Elettronica™ EU-C Atex Technical Balances
Designed for use in hazardous environments with the risk of explosion, these scales are classified as ZONE 2. Gibertini Elettronica™ EU-C Atex Technical Balances are built with method of protection nAL for gas group IIC and temperature class T4 (135°C) according to regulation EEC 94/9.
Brand: Gibertini Elettronica™ EU-C7500 PTX
Additional Details : Weight : 4.50000kg
Description
- LCD display with small-size decimal digits, for easier and more immediate reading
- Waterproof and acid resistant membrane keyboard. Easy to use with the ON/OFF, TARE, MODE and PRINT keys (RANGE/PRINT for DR model)
- Indication of the reached stable weight
- Bar-graph indicator of dosage and remaining capacity of the balance
- Parameters configurable by menu: reading in g (grams), lb (pound), oz (ounce), ct (carats), pcs (pieces), % (percentage)
- Automatic end of range calibration (with external mass supplied on request)
- Memory of the current weight in case of power failure
- Enhanced anti-shock system
- Metallic body with epoxide protection
- Load cell nearly unbreakable and without transportation problems
- Data output: I/O RS232 adjustable
- 1, 2 and 5kg masses in class F1 for calibration and special executions available upon request
Specifications
Classified as ZONE 2. Built with method of protect | |
0.1 g | |
±0.3 g | |
2 sec. | |
±0.3 | |
Technical Balance | |
190 mm | |
240 VAC |
7.5 kg | |
Designed for use in hazardous environments with th | |
Metallic Body with Epoxide Protection | |
EU-C 7500 PTX | |
10°C to 40°C | |
355 x 215 x 120 mm | |
4.5 kg | |
240 VAC |