missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38237-100ul
This item is not returnable.
View return policy
Description
ETV7 Polyclonal antibody specifically detects ETV7 in Human samples. It is validated for ELISA,Western Blot
Specifications
| ETV7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ETS translocation variant 7, ets variant 7, ets variant gene 7 (TEL2 oncogene), TEL-2, TEL2 oncogene, TEL2ETS-related protein Tel2, TELBEts transcription factor TEL-2b, Tel-related Ets factor, transcription factor ets, transcription factor ETV7, Transcription factor Tel-2, TREF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human ETV7 (NP_057219.1).,, Sequence:, MQEGELAISPISPVAAMPPLGTHVQARCEAQINLLGEGGICKLPGRLRIQPALWSREDVLHWLRWAEQEYSLPCTAEHGFEMNGRALCILTKDDFRHRAPSSGDVLYELLQYIKTQRRALVCGPFFGGIFRLKTPTQHSPVPPEEVTGPSQMDTRRGHLLQPPDPGLTSNFGHLDDPGLARWTPGKEESLNLCHCAELGCRTQGVCSFPA | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51513 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction