missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV6/Tel Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03434-100UL
This item is not returnable.
View return policy
Description
ETV6/Tel Polyclonal specifically detects ETV6/Tel in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ETV6/Tel | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2120 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ETS translocation variant 6, ets variant 6, ets variant gene 6 (TEL oncogene), ETS-related protein Tel1, Tel, TEL/ABL, TEL1, TELTEL1 oncogene, transcription factor ETV6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ETV6/Tel (NP_001978.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction