missing translation for 'onlineSavingsMsg'
Learn More

ETV6/Tel Antibody [PE], Novus Biologicals™

Product Code. 30735266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735266 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735266 Supplier Novus Biologicals Supplier No. AF7945P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ETV6/Tel Polyclonal antibody specifically detects ETV6/Tel in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV6/Tel
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias ETS translocation variant 6, ets variant 6, ets variant gene 6 (TEL oncogene), ETS-related protein Tel1, Tel, TEL/ABL, TEL1, TELTEL1 oncogene, transcription factor ETV6
Host Species Sheep
Immunogen E. coli-derived recombinant human ETV6, Ser2-Asp111, Accession # P41212
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2120
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.