Learn More
ETV6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13182-1-AP-20UL
 View more versions of this product
Product Code. 16801274
  • £135.00 / 20µL
Description
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
Specifications
Specifications
ETV6
Polyclonal
Unconjugated
ETV6
ETS related protein Tel1, ETS translocation variant 6, ets variant 6, ETV6, TEL, TEL/ABL, TEL1, Transcription factor ETV6
Rabbit
Antigen Affinity Chromatography
RUO
14011, 2120, 312777
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P41212, P97360
Etv6
ETV6 Fusion Protein Ag3852
20 μL
Primary
Rat, Human, Mouse