Learn More
Invitrogen™ ETV6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590606
Description
Immunogen sequence: MSETPAQCSI KQERISYTPP ESPVPSYASS TPLHVPVPRA LRMEEDSIRL PAHLRLQPIY WSRDDVAQWL KWAENEFSLR PIDSNTFEMN GKALLLLTKE DFRYRSPHSG DVLYELLQHI LKQRKPRILF SPFFHPGNSI HTQPEVILHQ NHEEDNCVQR TPRPSVDNVH HNPPTIELLH RSRSPITTNH RPSPDPEQRP LRSPLDNMIR RLSPAERAQG PRPHQENNHQ ESYPLSVSPM ENNHCPASSE SHPKPSSPRQ ESTRVIQLMP SPIMHPLILN PRHSVDFKQS RLSEDGLHRE; Positive Samples: HepG2, Mouse lung, Mouse spleen, Rat testis; Cellular Location: Nucleus.
This gene encodes an ETS family transcription factor. The product of this gene contains two functional domains: a N-terminal pointed (PNT) domain that is involved in the protein-protein interactions with itself and other proteins, and a C-terminal DNA-binding domain. Gene knockout studies in mice suggest that it is required for hematopoiesis and Maintenance of the developing vascular network. This gene is known to be involved in a large number of chromosomal rearrangements associated with leukemia and congenital fibrosarcoma.
Specifications
| ETV6 | |
| Polyclonal | |
| Unconjugated | |
| Etv6 | |
| AW123102; AW557856; CGI-108; CSL4; Csl4p; ETS translocation variant 6; ETS variant 6; ets variant gene 6 (TEL oncogene); ETS-related protein Tel1; Etv6; hCsl4p; p13; RP11-452K12.9; SKI4; Ski4p; Tel; TEL/ABL; Tel1; TEL1 oncogene; THC5; Transcription factor ETV6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14011, 2120, 312777 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.04 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P41212, P97360 | |
| Etv6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ETV6 (NP_0019781). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.