missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV5/ERM Antibody (3H3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51614-0.025ml
This item is not returnable.
View return policy
Description
ETV5/ERM Monoclonal antibody specifically detects ETV5/ERM in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| ETV5/ERM | |
| Monoclonal | |
| Unconjugated | |
| Ascites | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 3H3 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| ERMEts-related protein ERM, ETS translocation variant 5, ets variant 5, ets variant gene 5 (ets-related molecule), ets-related molecule | |
| Purified recombinant fragment of human ETV5/ERM expressed in E. Coli. | |
| 0.025 mL | |
| Cancer | |
| 2119 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction