missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV3 Monoclonal Antibody (1C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524392
This item is not returnable.
View return policy
Description
Peptide Sequence: MKAGCSIVEK PEGGGGYQFP DWAYKTESSP GSRQIQLWHF ILELLQKEEF RHVIAWQQGE YGEFVIKDPD EVARLWGRRK CKPQMNYDKL SRALRYYYNK RILHKTKGKR FTYKFNFNKL VMPNYPFINI RSSGKIQTLL VGN.
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
Specifications
| ETV3 | |
| Monoclonal | |
| Unconjugated | |
| ETV3 | |
| AI414410; bA110J1.4; ETS domain transcriptional repressor PE1; ETS translocation variant 3; ETS variant 3; ets variant gene 3; ets variant gene 3, ETS family transcriptional repressor; ETS-domain protein; ETS-domain transcriptional repressor; Etv3; Mets; Mitogenic Ets transcriptional suppressor; mitogenic Ets transcriptional suppressor METS; Pe1; PE-1; SH2 domain containing phosphatase anchor protein 1; Spap1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2117 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 1C11 | |
| PBS with no preservative; pH 7.4 | |
| P41162 | |
| ETV3 | |
| ETV3 (AAH22868, 1 a.a. ~ 143 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction