missing translation for 'onlineSavingsMsg'
Learn More

ETV3 Antibody (1C11), Novus Biologicals™

Product Code. 18398217 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398217 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398217 Supplier Novus Biologicals Supplier No. H00002117M02100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ETV3 Monoclonal antibody specifically detects ETV3 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV3
Applications ELISA
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH22868
Gene Alias bA110J1.4, ETS domain transcriptional repressor PE1, ETS translocation variant 3, ets variant 3, ets variant gene 3, ETS family transcriptional repressor, METS, Mitogenic Ets transcriptional suppressor, PE-1ets variant gene 3, PE1FLJ79173
Host Species Mouse
Immunogen ETV3 (AAH22868, 1 a.a. ~ 143 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKAGCSIVEKPEGGGGYQFPDWAYKTESSPGSRQIQLWHFILELLQKEEFRHVIAWQQGEYGEFVIKDPDEVARLWGRRKCKPQMNYDKLSRALRYYYNKRILHKTKGKRFTYKFNFNKLVMPNYPFINIRSSGKIQTLLVGN
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2117
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.