missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV1 Antibody (CL15251) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
£321.00 - £459.00
Specifications
| Antigen | ETV1 |
|---|---|
| Clone | CL15251 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279851
|
Novus Biologicals
NBP3-43657-100ul |
100 μL |
£459.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279490
|
Novus Biologicals
NBP3-43657-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ETV1 Monoclonal antibody specifically detects ETV1 in Human samples. It is validated for Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| ETV1 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 2115 | |
| IgG1 | |
| Protein A purified |
| CL15251 | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp781L0674, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534 | |
| This antibody was generated using a synthetic peptide of P50549, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title