missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV1 Antibody (1C8B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28868
This item is not returnable.
View return policy
Description
ETV1 Monoclonal specifically detects ETV1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ETV1 | |
| Monoclonal | |
| Unconjugated | |
| P50549 | |
| ETV1 | |
| Purified recombinant fragment of ETV1(aa1-191) expressed in E. Coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 2115 | |
| Human | |
| Ascites |
| Western Blot, ELISA | |
| 1C8B6 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| DKFZp781L0674, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for ETV1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction