Learn More
Cytiva Ettan™ IPGphor™ 3 Control Software
Product Code. 10177584 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
IEF System
Unit Size:
Each
Product Code. 10177584
Brand: Cytiva 11003623
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Monitors voltage, current and volt-hours of run.
Any number of protocols can be stored on a connected PC running Ettan IPGphor 3 control software and uploaded to the instrument instantly.
Control Software
  • Generates professional graphical reports with large user interface for programming and editing
  • Create, save, and edit protocols and also pause during run to edit protocol in real time
  • Control up to four Ettan IPGphor 3 IEF units simultaneously, each running different set of run parameters
  • Enables web browser remote monitoring of PC control unit
  • Stored log files of previous runs can be opened and viewed and status reports can be generated along with instantaneous run condition reports on request
  • Microsoft excel and other log files can be exported
  • Exports can be created, saved, printed, and exported
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) IEF System
Type Control Software
Includes Monitors voltage, current and volt-hours of run