missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Ettan DALTsix Electrophoresis Unit
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
2-D Electrophoresis
Unit Size:
Each
Description
- Runs up to six lab-cast or precast gels (26 × 20cm) simultaneously
- Use with 18- or 24 cm Immobiline DryStrip gels
- Stable binding with no or very low dissociation of captured antibodies
- Simple-to-use 1mm or 1.5mm pre-assembled hinged gel cassettes
- Innovative design simplifies loading and unloading
- Built-in ceramic heat exchanger and buffer circulation pump ensure efficient cooling
Specifications
Specifications
| For Use With (Application) | large format and medium throughput 2-D electrophoresis |
| For Use With (Equipment) | 2-D Electrophoresis |
| Type | Ettan DALTsix Electrophoresis Unit |
| No. of Gels | 6 |
| Volume (Metric) Buffer | 4.5 L (anode) 0.8 L (cathode) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction