Learn More
Descrizione
After the second-dimension run is complete, the large-format gels can be stained using DALT staining tray set and PlusOne silver staining kit, Protein. This combination ensures reliable detection with high sensitivity.The set includes two stainless steel trays for staining solutions; one perforated stainless steel tray that seats within the staining trays; and a transparent plastic cover. The perforated insert supports and restrains gels for transfer between staining trays while allowing staining solutions to drain rapidly.
- For Use With: Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large vertical System
Kit Contains:
- 2 x Tray for staining solutions (stainless steel)
- 1 x Perforated tray that seats within the staining trays (23x42 cm, stainless steel)
- 1 x Cover (transparent plastic)
- Instructions
Specifica
Specifica
| For Use With (Application) | Reproducible high resolution, high throughput 2D electrophoresis |
| For Use With (Equipment) | Ettan DALTtwelve Large Vertical System and Ettan DALTsix Large Vertical System |
| Type | Staining Tray Set |
| Includes | 2 Tray for staining solutions, 1 Perforated tray that seats within the staining trays, 1 Plastic cover, instructions |
| Quantity | 1 Kit |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.