Learn More
Cytiva DALT Gel 12.5
Shop All Cytiva ProductsDescription
DALT gel 12.5 (26 x 20 cm) is a precast polyacrylamide gel bound to a plastic support film, which provides ease of handling and dimensional stability. The gels are barcoded for convenient sample and batch tracking. DALT gel 12.5 is used together with DALT buffer kit, which comprises concentrated buffers for running the gel, gel buffer for inserting the gel into dalt precast gel cassette, and sealing solution for application of the ipg strip. The buffer in the gel is part of a buffer system that gives longer shelf-life. Precast gel cassettes improve handling efficiency and throughput. Gels can be quickly loaded and unloaded with minimal hands-on time.
- Highest capacity electrophoresis unit with the largest precast gels available (26 x 20 cm)
- Stable, discontinuous buffer system in a precast, homogeneous gel features high batch-to-batch reproducibility
- Simple-to-use gel cassettes for gel loading, unloading, and storage
- Programmable power supply and temperature control ensure high reproducibility among runs
- System design minimizes buffer consumption
Specifications
Specifications
| Buffer | Buffer based on piperidinopropionamide (PPA) |
| Color | Colorless |
| Content And Storage | Shelf Life: 8 months; Storage: 2 to 8°C |
| Format | Solid |
| Quantity | 6 Pk. |
| Separation Range | 12,000 to 120,000 Mr |
| Dimensions (L x W x H) | 255 x 196 x 1 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.