Learn More
Invitrogen™ ETS2 Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA19543
Description
PA1-9543 detects ETS2 in human samples. PA1-9543 has been successfully used in Western blot procedures. By Western blot, PA1-9543 detects a 55 kDa protein representing ETS2. PA1-9543 immunogen is a purified recombinant ETS2.3 corresponding to the chromosome 21 gene (Human).
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.
Specifications
| ETS2 | |
| Polyclonal | |
| Unconjugated | |
| ETS2 | |
| AU022856; avian erythroblastosis virus E26 (v-ets) oncogene homolog 2; E26 avian leukemia oncogene 2, 3' domain; ETS proto-oncogene 2, transcription factor; Ets2; Ets-2; ETS2IT1; oncogene ETS-2; protein C-ets-2; v-ets avian erythroblastosis virus E2 oncogene homolog 2; v-ets avian erythroblastosis virus E26 oncogene homolog 2; v-ets erythroblastosis virus E26 oncogene homolog 2 | |
| Chicken | |
| IgY fraction | |
| RUO | |
| 2114 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P15036 | |
| ETS2 | |
| Purified recombinant ETS2.3 corresponding to the chromosome 21 gene (Human). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.