missing translation for 'onlineSavingsMsg'
Learn More

ETS2 Antibody (PCRP-ETS2-1D9), Allophycocyanin, Novus Biologicals™

Product Code. 18339764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339764 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339764 Supplier Novus Biologicals Supplier No. NBP314209APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ETS2 Monoclonal antibody specifically detects ETS2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETS2
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-ETS2-1D9
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian)
Host Species Mouse
Immunogen Recombinant full-length human ETS2 protein (Uniprot: P15036)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2114
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.