missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586282
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
ETS1 (c-Ets-1; v-ets erythroblastosis virus E26 oncogene homlog 1 (avian); ETS-1; EWSR2; p54), a 441 A. A protein, is a member of DNA-binding ETS protein family. It is a multifunctional protein consisting of an N-terminal pointed domain, a central transactivation domain and 2 auto inhibitory domains flanked on both sides of conserved C-terminal Ets domain. It interacts with factors including PEBP2alphaA, p300/CBP and mediates transactivation of Osteopontin, Presenilin-1, Stromelysin and Collagenase. It is activated by Ras/MAPK signaling and its expression is up-regulated in resting T-cells. Its activity is modulated by various factors including AP-1, Sp-1, c-myb and MafB. Sp100 activates ETS1 in nuclear bodies. It interacts with EAP1/Daxx and causes repression of MMP1 and BCL2 transactivation. It interacts with STAT6 and modulates Socs-1 expression induced by IL-4. It interacts with GATA3 and positively regulates Tax-1 dependent IL-5 expression.
Specifications
| ETS1 | |
| Polyclonal | |
| Unconjugated | |
| ETS1 | |
| AI196000; AI448617; Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1; avian erythroblastosis virus E26 oncogene 1; c-ets protein; c-ets1; c-ets-1; c-ets-1 oncogene; c-ets-1 protein; c-ets-alpha; c-ets-beta; D230050P06; E26 avian leukemia oncogene 1, 5' domain; Ets avian erythroblastosis virus E2 oncogene homolog 1 (tumor progression locus 1); ets protein; ETS proto-oncogene 1, transcription factor; Ets1; ETS-1; ETS-1A; ETS-1B; Ets-1Delta(III-VI); Etsoncb; EWSR2; FLJ10768; oncoprotein; p54; p54 transcription factor; protein C-ets-1; TNFAIP3 interacting protein 1; TNIP1; Tpl1; transforming protein p54/c-ets-1; transforming protein p68/c-ets-1; tumor progression locus 1; unnamed protein product; v-ets avian erythroblastosis virus E2 oncogene homolog 1; v-ets avian erythroblastosis virus E26 oncogene homolog 1; v-ets erythroblastosis virus E26 oncogene homolog 1; virion-associated nuclear-shuttling protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2113, 23871, 24356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14921, P27577, P41156 | |
| ETS1 | |
| Synthetic peptide, corresponding to the N-terminus of Human ETS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction