missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETS1 Monoclonal Antibody (1601CT512.35.61)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537698

Product Code. 17127288

  • £454.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ETS1 Monoclonal Antibody for Western Blot, Flow

ETS1 (c-Ets-1; v-ets erythroblastosis virus E26 oncogene homlog 1 (avian); ETS-1; EWSR2; p54), a 441 A. A protein, is a member of DNA-binding ETS protein family. It is a multifunctional protein consisting of an N-terminal pointed domain, a central transactivation domain and 2 auto inhibitory domains flanked on both sides of conserved C-terminal Ets domain. It interacts with factors including PEBP2alphaA, p300/CBP and mediates transactivation of Osteopontin, Presenilin-1, Stromelysin and Collagenase. It is activated by Ras/MAPK signaling and its expression is up-regulated in resting T-cells. Its activity is modulated by various factors including AP-1, Sp-1, c-myb and MafB. Sp100 activates ETS1 in nuclear bodies. It interacts with EAP1/Daxx and causes repression of MMP1 and BCL2 transactivation. It interacts with STAT6 and modulates Socs-1 expression induced by IL-4. It interacts with GATA3 and positively regulates Tax-1 dependent IL-5 expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ETS1
Monoclonal
Unconjugated
ETS1
AI196000; AI448617; Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1; avian erythroblastosis virus E26 oncogene 1; c-ets protein; c-ets1; c-ets-1; c-ets-1 oncogene; c-ets-1 protein; c-ets-alpha; c-ets-beta; D230050P06; E26 avian leukemia oncogene 1, 5' domain; Ets avian erythroblastosis virus E2 oncogene homolog 1 (tumor progression locus 1); ets protein; ETS proto-oncogene 1, transcription factor; Ets1; ETS-1; ETS-1A; ETS-1B; Ets-1Delta(III-VI); Etsoncb; EWSR2; FLJ10768; oncoprotein; p54; p54 transcription factor; protein C-ets-1; TNFAIP3 interacting protein 1; TNIP1; Tpl1; transforming protein p54/c-ets-1; transforming protein p68/c-ets-1; tumor progression locus 1; unnamed protein product; v-ets avian erythroblastosis virus E2 oncogene homolog 1; v-ets avian erythroblastosis virus E26 oncogene homolog 1; v-ets erythroblastosis virus E26 oncogene homolog 1; virion-associated nuclear-shuttling protein
Mouse
Protein G
RUO
2113
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
1601CT512.35.61
PBS with 0.09% sodium azide; pH 7.4
P14921
ETS1
recombinant protein of human ETS1.
200 μL
Primary
Human
Antibody
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.