missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 associated protein II Antibody (TDP2/1258), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50071MFV610
This item is not returnable.
View return policy
Description
ETS1 associated protein II Monoclonal antibody specifically detects ETS1 associated protein II in Human samples. It is validated for Flow Cytometry
Specifications
| ETS1 associated protein II | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| TDP2/1258 | |
| Flow Cytometry | |
| AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2 | |
| Recombinant full-length human ETS1 associated protein II protein (Uniprot: O95551) | |
| 0.1 mL | |
| Signal Transduction, Transcription Factors and Regulators | |
| 51567 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction