missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ets-1 Antibody (ETS1/1801), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54514JF549
This item is not returnable.
View return policy
Description
Ets-1 Monoclonal antibody specifically detects Ets-1 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| Ets-1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, ETS-1, EWSR2, FLJ10768, p54, protein C-ets-1, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1, v-ets erythroblastosis virus E26 oncogene homolog 1 (avian) | |
| Recombinant human Ets-1 fragment (around aa137-230) (exact sequence is proprietary) (Uniprot: P14921) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Transcription Factors and Regulators | |
| 2113 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF | |
| ETS1/1801 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction