missing translation for 'onlineSavingsMsg'
Learn More

Ets-1 Antibody, Biotin, Novus Biologicals™

Product Code. 18398953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398953 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398953 Supplier Novus Biologicals Supplier No. NBP298794B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ets-1 Polyclonal specifically detects Ets-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ets-1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, ETS-1, EWSR2, FLJ10768, p54, protein C-ets-1, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1, v-ets erythroblastosis virus E26 oncogene homolog 1 (avian)
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Ets-1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2113
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.