missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ets-1 Antibody (8A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22169-0.025ml
This item is not returnable.
View return policy
Description
Ets-1 Monoclonal specifically detects Ets-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Ets-1 | |
| Monoclonal | |
| Unconjugated | |
| Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, ETS-1, EWSR2, FLJ10768, p54, protein C-ets-1, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1, v-ets erythroblastosis virus E26 oncogene homolog 1 (avian) | |
| Mouse | |
| 50.4 kDa | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 2113 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8A8 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| ETS1 | |
| Purified recombinant fragment of human ETS1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction