missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etripamil, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Etripamil (MSP-2017) is a short-acting L-type calcium-channel antagonist, can be used for the research of Paroxysmal Supraventricular Tachycardia (PSVT). Etripamil (MSP-2017) slows atrioventricular nodal conduction and prolongs atrioventricular nodal refractory periods by inhibiting calcium ion influx through the calcium slow channels in the atrioventricular node cells.
Specifications
Specifications
| Chemical Name or Material | Etripamil |
| CAS | 1593673-23-4 |
| Color | Reddish Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C27H36N2O4 |
| Quantity | 10 mg |
| Synonym | MSP-2017 (-)-MSP-2017 |
| Solubility Information | Ethanol : 120 mg/mL (265.14 mM; Need ultrasonic) |
| SMILES | O=C(OC)C1=CC=CC(CCN(CCC[C@](C2=CC=C(OC)C(OC)=C2)(C#N)C(C)C)C)=C1 |
| Molecular Weight (g/mol) | 452.59 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction