missing translation for 'onlineSavingsMsg'
Learn More

Etripamil, MedChemExpress

Product Code. 16443447
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443447

Brand: Medchem Express HY17611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etripamil (MSP-2017) is a short-acting L-type calcium-channel antagonist, can be used for the research of Paroxysmal Supraventricular Tachycardia (PSVT). Etripamil (MSP-2017) slows atrioventricular nodal conduction and prolongs atrioventricular nodal refractory periods by inhibiting calcium ion influx through the calcium slow channels in the atrioventricular node cells.

Etripamil (MSP-2017) is a short-acting L-type calcium-channel antagonist, can be used for the research of Paroxysmal Supraventricular Tachycardia (PSVT). Etripamil (MSP-2017) slows atrioventricular nodal conduction and prolongs atrioventricular nodal refractory periods by inhibiting calcium ion influx through the calcium slow channels in the atrioventricular node cells.

Specifications

Chemical Name or Material Etripamil
CAS 1593673-23-4
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C27H36N2O4
Quantity 50 mg
Synonym MSP-2017 (-)-MSP-2017
Solubility Information Ethanol : 120 mg/mL (265.14 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=CC(CCN(CCC[C@](C2=CC=C(OC)C(OC)=C2)(C#N)C(C)C)C)=C1
Molecular Weight (g/mol) 452.59
Formula Weight 452.59
Percent Purity 98.68%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.