missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etretinate, Mikromol™
Description
Etretinate
Specifications
Specifications
| Chemical Name or Material | Etretinate |
| CAS | 54350-48-0 |
| Analyte or Component Names | Etretinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H30 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,4,6,8-Nonatetraenoic acid, 9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-, ethyl ester, (2E,4E,6E,8E)-, 2,4,6,8-Nonatetraenoic acid, 9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-, ethyl ester, (all-E)-, Ethyl all-trans-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoate, Ethyl etrinoate, Etretinate, Ro 10-9359, Tegison, Tigason, Tigasone, Acitretin Related Compound B, Acitretin Impurity B |
| InChI Formula | InChI=1S/C23H30O3/c1-8-26-23(24)14-17(3)11-9-10-16(2)12-13-21-18(4)15-22(25-7)20(6)19(21)5/h9-15H,8H2,1-7H3/b11-9+,13-12+,16-10+,17-14+ |
| SMILES | CCOC(=O)\C=C(/C)\C=C\C=C(/C)\C=C\c1c(C)cc(OC)c(C)c1C |
| IUPAC Name | ethyl (2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?