missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epiberberine chloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Epiberberine chloride has antioxidant activity, with peroxynitrite ONOO- scavenging effect (IC50, 16.83 μM), and may protect against Alzheimer disease. Epiberberine chloride inhibits the early stage of differentiation of 3T3-L1 preadipocytes, downregulates the Raf/MEK1/2/ERK1/2 and AMPKα/Akt pathways. Epiberberine has the potential effect in the research of diabetic disease.
Specifications
Specifications
| Chemical Name or Material | Epiberberine chloride |
| CAS | 889665-86-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H18ClNO4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (67.24 mM; Need ultrasonic) |
| SMILES | COC1=C(OC)C=C2C(CC[N+]3=C2C=C(C=C4)C(C5=C4OCO5)=C3)=C1.[Cl-] |
| Molecular Weight (g/mol) | 371.81 |
| Formula Weight | 371.81 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction