missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ETP 45835 dihydrochloride
GREENER_CHOICE

Product Code. 18724739
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724739 10 mg 10mg
18700268 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18724739 Supplier Tocris Bioscience™ Supplier No. 5183/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mnk1 and Mnk2 inhibitor

ETP 45835 dihydrochloride is a Mnk2 and Mnk1 inhibitor (IC50 values are 575 and 646 nM respectively). Exhibits selectivity against a panel of 24 protein kinases, including those upstream of Mnk1/2.
TRUSTED_SUSTAINABILITY

Specifications

Purity >99%
Quantity 10 mg
Product Type ETP 45835 dihydrochloride
Molecular Weight (g/mol) 301.21
Inhibitors ETP 45835 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.