missing translation for 'onlineSavingsMsg'
Learn More

Etosalamide, MedChemExpress

Product Code. 16438648
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438648 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438648 Supplier Medchem Express Supplier No. HYB1015
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Etosalamide (Ethosalamide) is an antipyretic and analgesic agent. Etosalamide has anti-inflammatory activity and can be used for allergic disease research.

Etosalamide (Ethosalamide) is an antipyretic and analgesic agent. Etosalamide has anti-inflammatory activity and can be used for allergic disease research.

Specifications

Chemical Name or Material Etosalamide
CAS 15302-15-5
Color White
Purity Grade Notes Research
Molecular Formula C11H15NO3
Quantity 10 mg
Synonym Ethosalamide
Solubility Information DMSO : 300 mg/mL (1433.76 mM; Need ultrasonic)
SMILES O=C(N)C1=CC=CC=C1OCCOCC
Molecular Weight (g/mol) 209.24
Formula Weight 209.24
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.