missing translation for 'onlineSavingsMsg'
Learn More

Etoricoxib-d4, MedChemExpress

Product Code. p-200096897
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16499037

Brand: Medchem Express HY15321S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etoricoxib D4 (MK-0663 D4) is a deuterium labeled Etoricoxib. Etoricoxib is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.

Etoricoxib D4 (MK-0663 D4) is a deuterium labeled Etoricoxib. Etoricoxib is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.

Specifications

Chemical Name or Material Etoricoxib-d4
CAS 1131345-14-6
Purity Grade Notes Research
Physical Form Powder
Molecular Formula C18H11D4ClN2O2S
Color Light Nude
Synonym MK-0663-d4 L-791456-d4
SMILES ClC(C=N1)=CC(C2=C([2H])C([2H])=C(S(C)(=O)=O)C([2H])=C2[2H])=C1C3=CN=C(C)C=C3
Molecular Weight (g/mol) 362.87
Quantity 5 mg
Formula Weight 362.87
Percent Purity 99.35%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.